Learn More
Aviva Systems Biology Corp Smpdl3a Antibody - N-terminal region (ARP52244_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52244_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Smpdl3a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 0610010C24Rik, AI529588, ASM3A, ASML3, ASML3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-670