Learn More
Cayman Chemical SMPDL3AhumnrecombInt100ug
SDP
Catalog No. 502944009
Encompass_Preferred
Source: Recombinant C-Terminal His-tagged SMPDL3A expressed in HEK293 cells • Amino acids: 23-453 • MW: 50.3 kDa
Catalog No. 50-294-4009 Supplier Cayman Chemical Supplier No. 40468100 UG
Source: Recombinant C-Terminal His-tagged SMPDL3A expressed in HEK293 cells • Amino acids: 23-453 • MW: 50.3 kDa