Learn More
Aviva Systems Biology Corp SMPDL3B Antibody - N-terminal region (ARP33875_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMPDL3B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SMPDL3B. Alternate names: ASML3BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55183