Learn More
Medchemexpress LLC SMPH crosslinker | 367927-39-7 | MFCD01863460 | 95.0% | 379.36 | C17H21N3O7 | 250MG
SDP
Catalog No. 5000236937
Encompass_Preferred
SMPH crosslinker is an alkyl/ether-based bifunctional linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester, used to connect ligands and create conjugates in synthesis workflows such as PROTAC construction and antibody-drug conjugation. The material is supplied as a white to off-white solid with formula C17H21N3O7 and a molecular weight of 379.36.
Catalog No. 50-002-36937 Supplier Medchemexpress LLC Supplier No. HY124318250MG
SMPH crosslinker is an alkyl/ether-based bifunctional linker containing a maleimide and an N-hydroxysuccinimide (NHS) ester, used to connect ligands and create conjugates in synthesis workflows such as PROTAC construction and antibody-drug conjugation. The material is supplied as a white to off-white solid with formula C17H21N3O7 and a molecular weight of 379.36.