Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Sample Cell: 1" Square Glass 25mL, matched pair

Catalog No. 50465396 Shop All Hach Company Products
Encompass

Sample cell, 1 inch 25 mL. Matched pair for use with the DR/3000.

Catalog No. 50-465-396 Supplier Hach Company Supplier No. 1993500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sample cell, 1 inch 25 mL. Matched pair for use with the DR/3000.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.