Learn More
Research Products International Corp Sample Head Set for Vortex-Genie 2, Multiple
SDP
Catalog No. 502229782
Encompass
The Multiple Sample Starter Set can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse. It consists of a 6 inch (152mm) Platform, Microtube Foam Insert and Microplate Foam Insert . Each insert fits snugly into the 6 inch Platform attachment to accommodate either 60 microtubes or one standard footprint microplate, respectively.
Catalog No. 50-222-9782 Supplier Research Products International Corp Supplier No. 155301
The Multiple Sample Starter Set can be used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse. It consists of a 6 inch (152mm) Platform, Microtube Foam Insert and Microplate Foam Insert . Each insert fits snugly into the 6 inch Platform attachment to accommodate either 60 microtubes or one standard footprint microplate, respectively.