Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Use Gamma Irradiated Sampling Bottle Assembly, PETG Bottle; 125 mL
Catalog No. NC1813029
Encompass
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 125 mLSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 125 mL bottle
Catalog No. NC1813029 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564420
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 125 mLSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 125 mL bottle