Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Use Gamma Irradiated Sampling Bottle Assembly, PETG Bottle; 1 L
Catalog No. NC1813033
Encompass
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 1 LSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 1 L 1000 mL
This item is not returnable. View return policy
Catalog No. NC1813033 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564426
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 1 LSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 1 L 1000 mL