Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Use Gamma Irradiated Sampling Bottle Assembly, PETG Bottle; 2 L
Catalog No. NC1813035
Encompass
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 2 LSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 2 L 2000 mL
Catalog No. NC1813035 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564428
Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 2 LSafely transfer, sample, and store biopharmaceutical products and reagentsSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are gamma irradiatedeliminates labor and validation costs associated with assembly and cleaningDesigned with 2 L 2000 mL