Learn More
Aviva Systems Biology Corp SMR3A Antibody - middle region (ARP42363_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42363_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMR3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMR3A. Alternate names: P-B1, PBI, PRL5, PROL5ApplicationWB. Species reactivity: Cow, Horse, Human, Rat
Catalog No. 50-014-58199