Learn More
Sino Biological SMS 293-SUPI Expression Medium Supplement 100 mL
Catalog No. 5000696222
Encompass_Preferred
SMS 293-SUPI is a protein-free, serum-free liquid supplement that is specifically used to meet the nutritional requirements of HEK293(such as 293-F, 293H, etc.) suspension cells at high-density growth and late transient transfection. This supplement contains high concentrations of essential nutrients, such as amino acids and inorganic salts, required for cell growth?without L-Glutamine.
Catalog No. 50-006-96222 Supplier Sino Biological Supplier No. M293SUPI100
SMS 293-SUPI is a protein-free, serum-free liquid supplement that is specifically used to meet the nutritional requirements of HEK293(such as 293-F, 293H, etc.) suspension cells at high-density growth and late transient transfection. This supplement contains high concentrations of essential nutrients, such as amino acids and inorganic salts, required for cell growth?without L-Glutamine.