Learn More
Aviva Systems Biology Corp Sms Antibody - middle region (ARP63041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sms. It was validated on Western Blot. . Alternate names: AI427066, Gy, SpmST, gyro, SPMSYApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-852