Learn More
MSC Ansell AlphaTec 1500 Coveralls, SMS Fabric, Small, 25/Case
Catalog No. 5000686493
Encompass
These disposable protective coveralls are constructed from breathable SMS nonwoven fabric to provide a reliable barrier against hazardous particles and light liquid sprays. The suit is designed to minimize heat stress while ensuring comfort and coverage in various industrial applications.
  • Filters 100% of particles over 3 microns
  • Includes attached three-piece elasticated hood, boots, and elasticated wrists and ankles
  • Features a two-way front zipper with a resealable storm flap
  • Silicone-free design for compatibility with spray-painting
  • Optimized body fit for improved movement
  • Certified anti-static properties
  • Suitable for asbestos removal, powder handling, and general maintenance
  • Standards met: CE Category III, Type 5, Type 6, EN 1073-2, EN ISO 13688:2013, ASTM F1001, and ISO 6529
Catalog No. 50-006-86493 Supplier MSC Supplier No. 70222237
These disposable protective coveralls are constructed from breathable SMS nonwoven fabric to provide a reliable barrier against hazardous particles and light liquid sprays. The suit is designed to minimize heat stress while ensuring comfort and coverage in various industrial applications.
  • Filters 100% of particles over 3 microns
  • Includes attached three-piece elasticated hood, boots, and elasticated wrists and ankles
  • Features a two-way front zipper with a resealable storm flap
  • Silicone-free design for compatibility with spray-painting
  • Optimized body fit for improved movement
  • Certified anti-static properties
  • Suitable for asbestos removal, powder handling, and general maintenance
  • Standards met: CE Category III, Type 5, Type 6, EN 1073-2, EN ISO 13688:2013, ASTM F1001, and ISO 6529