Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety SMS Disposable Anti-Static Frocks, No Pockets
SDP

Catalog No. 19813032
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-032 Small
19-813-033 Medium
19-813-034 Large
19-813-035 X-Large
19-813-036 2X-Large
19-813-037 3X-Large
19-813-038 4X-Large
19-813-039 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-813-032 Supplier Keystone Safety Supplier No. CEF0WKSMSSMWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Knee-length frocks constructed of SMS composite fabric feature knit wrists and elastic thumb loops. They provide particulate and liquid barrier suitable for ISO Class 6 cleanrooms.

  • Made from SMS composite fabric
  • Disposable, without pockets
  • Highly breathable and comfortable
  • ISO Class 6 cleanroom compatible
  • Low shedding and anti-static to protect sensitive products
  • Provide particulate barrier and liquid repellant
  • Ideal for food processing, laboratories, biohazards, medical, metal surface treating, pharmaceutical
  • Knit cuffs with elastic thumb loops, six snap closure, knit collar, extra-long knee-length, hanger loop

Specifications

Size Small
Material SMS
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Knit, Elastic Thumb Loop
Collar Type Knit
Latex Free Yes
Sleeve Style Long Sleeve
Cleanroom Class Class 1000 (ISO 6)
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.