Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety SMS Lab Coat
SDP

Catalog No. 19070131
Change view
Click to view available options
Size:
4X-Large
Closure Type:
Zipper
Cuff Style:
Open
No. of Pockets:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Closure Type Cuff Style No. of Pockets
19-070-131 4X-Large Zipper Open 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-070-131 Supplier Keystone Safety Supplier No. LC3BKSMS4XLZIP
This item is not returnable. View return policy

Keystone SMS material is soft and durable, which makes it easy to work with. It offers excellence barrier protection while keeping the worker cool, comfortable and does not sacrifice mobility and comfort.

  • Non linting
  • 100% latex free and flame resistant
  • Individually inspected for stitching, sizing, material and workmanship
  • Ideal for class 1000 environmental and above
  • Commonly used in electronic, medical device and pharmaceutical industries

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Color Blue
Closure Type Zipper
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Latex Free Yes
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Construction SMS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.