Learn More
Fisherbrand™ SMS Lab Jacket Cranberry
Disposable lab jacket with anti-static coating. Knit wrist cuffs provide comfortable and secure fit.
Supplier:  Fisherbrand™ 179375
Catalog No. 17-103-079
Only null left
Add to Cart
Description
Description
Single-use lab coat, Made of 40gsm SMS anti-static coating:
  • One outside left breast pocket
  • Two outside lower pockets
  • Knit collar
  • Front snap closure
  • Knit cuffs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.