Learn More
International Enviroguard™ SMS 2001 Shoe Covers
Catalog No. 19472795 Shop All International Enviroguard Systems Products
Change view
Click to view available options
SMS Shoe Covers; Size: X-Large; Closure Type: Elastic; Material: SMS; Color: White
Catalog No.
19-472-795 SMS Shoe Covers; Size: X-Large; Closure Type: Elastic; Material: SMS; Color: White
1 options
Catalog No. 19-472-795 Supplier International Enviroguard™ Supplier No. 2001
Add to Cart
Add to Cart
Disposable SMS shoe covers keep floors and walkways clean from contaminants found on the soles of shoes.
SMS shoe covers provide the breathability feet need to stay cool for all-day comfort. These durable and lightweight shoe covers are made of three-layer spunbond meltblown spunbond (SMS) fabric that helps keep floors and walkways clean from debris that gets tracked around. These disposable shoe covers also help protect your shoes from damage or buildup of debris like mud and dirt. Keep dust, dirt, and other particulates from dirtying your workplace and surrounding areas with these cost-effective shoe covers.
  • Possible applications include construction, food processing, pharmaceuticals, environmental cleanup, beauty, contract and installation work, movers, and more.
  • Large opening at top makes it easy to slip over your shoes
  • Elastic closure top for a secure fit that helps keep shoe covers in place
  • Fits up to a men's size 12 shoe or size 11 boot
  • Can be worn in indoor and outdoor environments
Specifications
Size X-Large
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Standard
Resistance Dry Particulates
Product Type Controlled Environments Shoe Covers