Learn More
Medchemexpress LLC Sms2-in-1 | 2098890-15-2 | MFCD31813610 | 98.0% | 681.67 g·mol⁻¹ | C34H37F6N3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1020415MG
Encompass_Preferred
SMS2-IN-1 is a potent, highly selective small-molecule inhibitor of sphingomyelin synthase 2 (SMS2) for use in biochemical and cellular research. It demonstrates low-nanomolar activity against SMS2 with strong selectivity over related isoforms, is supplied as a solid for dissolution in DMSO, and is offered in multiple research-scale pack sizes.
  • Potent SMS2 inhibition with IC50 6.5 nM and Kd 37 nM.
  • High selectivity over SMS1 (≈150-fold).
  • High purity (98.04%).
  • Solid, white to off-white appearance; soluble in DMSO.
  • Available in multiple small-scale pack sizes for research use.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-47540