Learn More
Medchemexpress LLC SMS2-IN-2 | 2241838-28-6 | 98.0% | 369.78 | 50 MG
SDP
Catalog No. 5000437099
Encompass_Preferred
SMS2-IN-2 is a potent, highly selective, and orally active sphingomyelin synthase 2 (SMS2) inhibitor. It has shown anti-chronic inflammatory activity in animal models.
  • Potent and highly selective SMS2 inhibitor (IC50s of 100 nM for SMS2 and 56 μM for SMS1).
  • Orally active.
  • Exhibits anti-chronic inflammatory activity.
  • Reduces chronic inflammation in animal models of Type 2 diabetes mellitus.
  • Significantly decreases IL-6 and insulin levels in db/db mice.
Catalog No. 50-004-37099 Supplier Medchemexpress LLC Supplier No. HY11271350MG
SMS2-IN-2 is a potent, highly selective, and orally active sphingomyelin synthase 2 (SMS2) inhibitor. It has shown anti-chronic inflammatory activity in animal models.
  • Potent and highly selective SMS2 inhibitor (IC50s of 100 nM for SMS2 and 56 μM for SMS1).
  • Orally active.
  • Exhibits anti-chronic inflammatory activity.
  • Reduces chronic inflammation in animal models of Type 2 diabetes mellitus.
  • Significantly decreases IL-6 and insulin levels in db/db mice.