Learn More
Enzo Life Sciences S-Methyl-L-thiocitrulline . dihydrochloride (10mg). CAS: 209589-59-3
SDP
Supplier:  Enzo Life Sciences ALX106012M010
Encompass_Preferred
NOS inhibitor. Inhibitor of neuronal nitric oxide synthase (nNOS; NOS I). Formula: C7H15N3O2S . 2HCl. MW: 205.3 . 73.0. Purity: ≥97%. Appearance: White to off-white solid. Solubility: Soluble in water. Long Term Storage: +4°C. Handling: Protect from light and moisture. Packaged under inert gas. Hygroscopic.
Catalog No. 50-200-7739