Learn More
Enzo Life Sciences Smooth muscle actin monoclonal antibody (CGA7) (0.5ml)
SDP
Supplier:  Enzo Life Sciences ENZC34933
Encompass_Preferred
Clone: CGA7. Isotype: IgG2b. Immunogen: Partially purified chicken gizzard actin. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human Rabbit, Monkey, Chicken. Specificity: Recognizes actin isotypes α and γ of smooth muscle cells. This antibody labels leiomyosarcomas and leiomyomas but does not react with other neoplasms such as carcinoma or lymphoma. Except for myoepithelium, it is nonreactive with other mesenchymal and all epithelial cells. Applications: IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-3371