Learn More
Aviva Systems Biology Corp SMU1 Antibody - N-terminal region (ARP57160_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57160_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMU1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMU1. Alternate names: BWD, DKFZp761L0916, FLJ10805, FLJ10870, FLJ11970, MGC117363, RP11-54K16.3, SMU-1, fSAP57ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62096