Learn More
GA International Thermal-transfer, smudge and alcohol-proof resin ribbons 1.57" x 984' / 40mm x 300m White 1" / 25.4mm
Catalog No. NC1630690
Encompass_Preferred
Smudge and alcohol proof resin ribbon for thermal-transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No. NC1630690 Supplier GA International Supplier No. RR40X300C11IZ4WH
Smudge and alcohol proof resin ribbon for thermal-transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.