Learn More
Aviva Systems Biology Corp SMUG1 Antibody - middle region (ARP55013_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55013_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMUG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMUG1. Alternate names: FDG, HMUDG, MGC104370, UNG3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61396