Learn More
TARGETMOL CHEMICALS INC SMURF1-IN-A01 10MG
SDP
Catalog No. 502577322
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Smurf1-IN-A01 (A01) is a ubiquitin ligase Smad ubiquitination regulatory factor-1 inhibitor (kd 3.664 nM). It increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation. purity: 99%
Catalog No. 50-257-7322 Supplier TARGETMOL CHEMICALS INC Supplier No. T1690410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Smurf1-IN-A01 (A01) is a ubiquitin ligase Smad ubiquitination regulatory factor-1 inhibitor (kd 3.664 nM). It increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation. purity: 99%