Learn More
Medchemexpress LLC HY-110195 10mg Medchemexpress, Smurf1-IN-A01 CAS:1007647-73-5 Purity:>98%
SDP
Catalog No. 501968767
Encompass_Preferred
Medchemexpress, HY-110195 10mg Smurf1-IN-A01 CAS:1007647-73-5 Smurf1-IN-A01 (A01) is an ubiquitin ligase Smad ubiquitination regulatory factor-1 (Smurf1) inhibitor with a kd of 3.664 nM, which increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8767 Supplier Medchemexpress LLC Supplier No. HY11019510MG
Medchemexpress, HY-110195 10mg Smurf1-IN-A01 CAS:1007647-73-5 Smurf1-IN-A01 (A01) is an ubiquitin ligase Smad ubiquitination regulatory factor-1 (Smurf1) inhibitor with a kd of 3.664 nM, which increases BMP-2 responsiveness by inhibiting Smurf1-mediated Smad1/5 degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.