Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Smurf1 ligand 1 | 99.3% | 386.41 | 25 MG
SDP

Catalog No. 5000394291
Encompass_Preferred

Smurf1 ligand 1 is a PROTAC target protein ligand that can be used for synthesizing SMART1 (HY-W998345). It is for research use only.

  • Molecular weight: 386.41
  • Formula: C21H18N6O2
  • Appearance: Solid
  • Color: Light brown to brown
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)

Catalog No. 50-003-94291 Supplier Medchemexpress LLC Supplier No. 25MGHY169049
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Smurf1 ligand 1 is a PROTAC target protein ligand that can be used for synthesizing SMART1 (HY-W998345). It is for research use only.

  • Molecular weight: 386.41
  • Formula: C21H18N6O2
  • Appearance: Solid
  • Color: Light brown to brown
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.