Learn More
Aviva Systems Biology Corp SMYD1 Antibody - middle region (ARP50862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMYD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMYD1. Alternate names: BOP, ZMYND18, ZMYND22, KMT3DApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60455