Learn More
Aviva Systems Biology Corp SMYD2 Antibody - N-terminal region (ARP48568_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48568_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMYD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMYD2. Alternate names: HSKM-B, KMT3C, MGC119305, ZMYND14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59861