Learn More
Medchemexpress LLC SMYD2 protein, human (sf9, His) | >95.0% | 10 UG
SDP
Catalog No. 5000292380
Encompass_Preferred
Recombinant human SMYD2 protein expressed in Sf9 insect cells with an N-terminal His tag. Supplied as purified protein suitable for biochemical assays, enzymatic methylation studies, antibody validation, and structural biology. Manufacturer documentation such as a datasheet, certificate of analysis (COA), and safety data sheet (SDS) are referenced by the supplier.
  • Recombinant expression in Sf9 insect cells.
  • N-terminal His tag for purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Available in low microgram quantities for research use.
  • Datasheet, COA, and SDS referenced for quality and handling.
Catalog No. 50-002-92380 Supplier Medchemexpress LLC Supplier No. HYP7607910UG
Recombinant human SMYD2 protein expressed in Sf9 insect cells with an N-terminal His tag. Supplied as purified protein suitable for biochemical assays, enzymatic methylation studies, antibody validation, and structural biology. Manufacturer documentation such as a datasheet, certificate of analysis (COA), and safety data sheet (SDS) are referenced by the supplier.
  • Recombinant expression in Sf9 insect cells.
  • N-terminal His tag for purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Available in low microgram quantities for research use.
  • Datasheet, COA, and SDS referenced for quality and handling.