Learn More
Aviva Systems Biology Corp SMYD3 Antibody - C-terminal region (ARP50667_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50667_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMYD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SMYD3. Alternate names: FLJ21080, MGC104324, ZMYND1, ZNFN3A1, bA74P14.1, KMT3EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60406