Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SMYD3-IN-1 | 2095160-79-3 | 98.0% | 100 MG
SDP

Catalog No. 5000338160
Encompass_Preferred

SMYD3-IN-1 is an irreversible and selective inhibitor of SMYD3, with an IC50 of 11.7 nM. It is intended for research use only. This compound has a molecular weight of 507.02, a chemical formula of C28H31ClN4O3, and appears as an off-white to pink solid. It exhibits solubility of 100 mg/mL in DMSO.

  • Selective SMYD3 inhibitor with an IC50 of 11.7 nM
  • Molecular weight of 507.02
  • Chemical formula: C28H31ClN4O3
  • Appears as an off-white to pink solid
  • Solubility of 100 mg/mL in DMSO
  • GI50 of 1.04 μM in HepG2 cells for inhibition of SMYD3 in anchorage-independent colony formation
  • GI50 of 17.69 μM in HepG2 cells for inhibition of cell proliferation

Catalog No. 50-003-38160 Supplier Medchemexpress LLC Supplier No. HY128352100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMYD3-IN-1 is an irreversible and selective inhibitor of SMYD3, with an IC50 of 11.7 nM. It is intended for research use only. This compound has a molecular weight of 507.02, a chemical formula of C28H31ClN4O3, and appears as an off-white to pink solid. It exhibits solubility of 100 mg/mL in DMSO.

  • Selective SMYD3 inhibitor with an IC50 of 11.7 nM
  • Molecular weight of 507.02
  • Chemical formula: C28H31ClN4O3
  • Appears as an off-white to pink solid
  • Solubility of 100 mg/mL in DMSO
  • GI50 of 1.04 μM in HepG2 cells for inhibition of SMYD3 in anchorage-independent colony formation
  • GI50 of 17.69 μM in HepG2 cells for inhibition of cell proliferation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.