Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SMYD3-IN-1 | 2095160-79-3 | 98.0% | 50 MG
SDP

Catalog No. 5000340416
Encompass_Preferred

SMYD3-IN-1 (compound 29) is an irreversible and selective inhibitor of SMYD3 (SET and MYND domain containing 3) with an IC50 of 11.7 nM. This compound is intended for research use only.

  • Irreversible and selective inhibitor of SMYD3
  • IC50 of 11.7 nM
  • Molecular weight of 507.02
  • Chemical formula C28H31ClN4O3
  • Appearance as an off-white to pink solid
  • Soluble in DMSO at 100 mg/mL
  • Demonstrates cellular effects in HepG2 cells with GI50 values of 1.04 μM (colony formation) and 17.69 μM (cell proliferation)

Catalog No. 50-003-40416 Supplier Medchemexpress LLC Supplier No. HY12835250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMYD3-IN-1 (compound 29) is an irreversible and selective inhibitor of SMYD3 (SET and MYND domain containing 3) with an IC50 of 11.7 nM. This compound is intended for research use only.

  • Irreversible and selective inhibitor of SMYD3
  • IC50 of 11.7 nM
  • Molecular weight of 507.02
  • Chemical formula C28H31ClN4O3
  • Appearance as an off-white to pink solid
  • Soluble in DMSO at 100 mg/mL
  • Demonstrates cellular effects in HepG2 cells with GI50 values of 1.04 μM (colony formation) and 17.69 μM (cell proliferation)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.