Learn More
Aviva Systems Biology Corp SMYD4 Antibody - N-terminal region (ARP50738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMYD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMYD4. Alternate names: KIAA1936, ZMYND21ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60422