Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ SMZ 160 Stereo Microscopes

Catalog No. 01258012 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Base Style:
LED Stand
Plain Stand
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Base Style
01-258-012 Binocular Plain Stand
01-258-014 Trinocular Plain Stand
01-258-013 Binocular LED Stand
01-258-015 Trinocular LED Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-258-012 Supplier Motic™ Supplier No. 1100201300151
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This stereo zoom microscope comes with a 1:6.4 zoom range and an all metal body with classic illumination.

The SMZ-160 is ideal for educational purposes in high schools and universities, but is also great for industrial quality control.

  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6:1 zoom ratio
  • Diopter Adjustment: ±5

Specifications

Base Style Plain Stand
Magnification Power 10X
Objective Types Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
Head Style Binocular
Eyepiece WF 10X/20 mm
Working Distance 100 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.