Learn More
Motic SMZ171-TLED Stereo and Camera Bundle
Catalog No. 01257488 Shop All Motic Group Products
Change view
Click to view available options
Model:
SMZ-171-TLED w/ BMH4000X Camera Bundle
SMZ-171-TLED with BTI10 Camera Bundle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01-257-488 SMZ-171-TLED with BTI10 Camera Bundle
01-257-889 SMZ-171-TLED w/ BMH4000X Camera Bundle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-257-488 Supplier Motic Supplier No. 1100200601131
Add to Cart
Add to Cart
This item is not returnable. View return policy
The SMZ-171 is an advanced stereo microscope for biomedical work and industrial quality control.
  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6.7:1 Zoom Ratio
  • Diopter Adjustment: ±5
Specifications
Magnification Power 10X
Objective Types Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
Model SMZ-171-TLED with BTI10 Camera Bundle
Head Style Trinocular
Camera Type Moticam BTI10
Lighting Type LED
Eyepiece WF 10X/23 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov