Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SN-008 | 2249106-01-0 | 99.68% | C25H19FN2O3S | 1 ML
SDP

Catalog No. 5000354849
Encompass_Preferred

SN-008 is a less active analog of SN-011 and can be utilized as a negative control. It is intended for research use only.

  • Less active analog of SN-011
  • Can be utilized as a negative control
  • Intended for research use only
  • High purity of 99.68%
  • Molecular weight of 446.49
  • Appears as a white to off-white solid
  • Soluble in DMSO

Catalog No. 50-003-54849 Supplier Medchemexpress LLC Supplier No. HY14500910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SN-008 is a less active analog of SN-011 and can be utilized as a negative control. It is intended for research use only.

  • Less active analog of SN-011
  • Can be utilized as a negative control
  • Intended for research use only
  • High purity of 99.68%
  • Molecular weight of 446.49
  • Appears as a white to off-white solid
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.