Learn More
TARGETMOL CHEMICALS INC SN-008 5MG
SDP
Catalog No. 502576200
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SN-008 is a less active analog SN-011 which is a STING antagonist. SN-008 can be used as a negative control. purity: 96%
Catalog No. 50-257-6200 Supplier TARGETMOL CHEMICALS INC Supplier No. T95545MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SN-008 is a less active analog SN-011 which is a STING antagonist. SN-008 can be used as a negative control. purity: 96%