Learn More
Medchemexpress LLC SN-011 | 2249435-90-1 | 99.9% | C25H19FN2O4S | 1 ML
SDP
Catalog No. 5000355737
Encompass_Preferred
SN-011 is a potent and selective inhibitor of both mouse and human STING signaling, with an IC50 of 76 nM. It functions by competing with cyclic dinucleotide (CDN) for the binding pocket of the STING dimer, thereby blocking CDN binding and subsequent STING activation. This compound is intended for research into STING-driven autoimmune and inflammatory diseases.
  • Potent and selective STING inhibitor
  • Inhibits STING signaling with an IC50 of 76 nM
  • Blocks CDN binding and STING activation
  • Supports research on STING-driven autoimmune and inflammatory diseases
Catalog No. 50-003-55737 Supplier Medchemexpress LLC Supplier No. HY14501010MM/1ML
SN-011 is a potent and selective inhibitor of both mouse and human STING signaling, with an IC50 of 76 nM. It functions by competing with cyclic dinucleotide (CDN) for the binding pocket of the STING dimer, thereby blocking CDN binding and subsequent STING activation. This compound is intended for research into STING-driven autoimmune and inflammatory diseases.
  • Potent and selective STING inhibitor
  • Inhibits STING signaling with an IC50 of 76 nM
  • Blocks CDN binding and STING activation
  • Supports research on STING-driven autoimmune and inflammatory diseases