Learn More
Medchemexpress LLC SN-011 | 2249435-90-1 | C25H19FN2O4S | 10MG
SDP
Catalog No. 5000260254
Encompass_Preferred
This small-molecule STING inhibitor blocks STING activation by competing with cyclic dinucleotides for the STING binding pocket, reducing downstream interferon signaling. It is reported to inhibit mouse and human STING with an IC50 of 76 nM and is intended for research into STING-driven immune signaling, inflammation, and autoimmunity.
  • Potent STING antagonist with reported IC50 of 76 nM.
  • Active against mouse and human STING.
  • Competitive inhibitor of cyclic dinucleotide binding.
  • Provided with analytical data including COA, H NMR, RP-HPLC, and MS.
  • Suitable for cell-based and biochemical studies of innate immune signaling.
Catalog No. 50-002-60254 Supplier Medchemexpress LLC Supplier No. HY14501010MG
This small-molecule STING inhibitor blocks STING activation by competing with cyclic dinucleotides for the STING binding pocket, reducing downstream interferon signaling. It is reported to inhibit mouse and human STING with an IC50 of 76 nM and is intended for research into STING-driven immune signaling, inflammation, and autoimmunity.
  • Potent STING antagonist with reported IC50 of 76 nM.
  • Active against mouse and human STING.
  • Competitive inhibitor of cyclic dinucleotide binding.
  • Provided with analytical data including COA, H NMR, RP-HPLC, and MS.
  • Suitable for cell-based and biochemical studies of innate immune signaling.