Learn More
Echelon Biosciences Research Labs SN-1-BIOTIN CARDIOLIPIN 10ug
SDP
Catalog No. 5082231
Encompass
sn-1-Biotin-Labeled Cardiolipin (Biotin-Cardiolipin) can be used in microplate and bead-based assays and affinity experiments. Cardiolipin (CL) is an important component in prokaryotic and in eukaryotic inner mitochondrial membrane. In humans, CL is made by cardiolipin synthase from phosphatidylglycerol and cytidinediphosphatediacylglycerol. Due to its four acyl chains and small head group, CL can organize into domains which can be protonsinks, fill cavities at protein interfaces, stabilize protein oligomers, and participate in high-curvature membrane regions. Autoantibodies to CL have been found in patients with anti-phospholipid syndrome and lupus. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-822-31 Supplier Echelon Biosciences Research Labs Supplier No. LC16B10UG
sn-1-Biotin-Labeled Cardiolipin (Biotin-Cardiolipin) can be used in microplate and bead-based assays and affinity experiments. Cardiolipin (CL) is an important component in prokaryotic and in eukaryotic inner mitochondrial membrane. In humans, CL is made by cardiolipin synthase from phosphatidylglycerol and cytidinediphosphatediacylglycerol. Due to its four acyl chains and small head group, CL can organize into domains which can be protonsinks, fill cavities at protein interfaces, stabilize protein oligomers, and participate in high-curvature membrane regions. Autoantibodies to CL have been found in patients with anti-phospholipid syndrome and lupus. This item is non returnable