Learn More
Medchemexpress LLC HY-13704 100mg Medchemexpress, SN-38 CAS:86639-52-3 Purity:>98%
SDP
Catalog No. 502029020
Encompass_Preferred
Medchemexpress, HY-13704 100mg SN-38 CAS:86639-52-3 SN-38 (NK012) is an active metabolite of the Topoisomerase I inhibitor Irinotecan. SN-38 (NK012) inhibits DNA and RNA synthesis with IC50s of 0.077 and 1.3 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9020 Supplier Medchemexpress LLC Supplier No. HY13704100MG
Medchemexpress, HY-13704 100mg SN-38 CAS:86639-52-3 SN-38 (NK012) is an active metabolite of the Topoisomerase I inhibitor Irinotecan. SN-38 (NK012) inhibits DNA and RNA synthesis with IC50s of 0.077 and 1.3 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.