Learn More
Selleck Chemical LLC SN-38 10mg 86639-52-3
SDP
Supplier:  Selleck Chemical LLC S490810MG
Encompass_Preferred
SN-38 is an active metabolite of CPT-11, inhibits DNA topoisomerase I, DNA synthesis and causes frequent DNA single-strand breaks. SN-38 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0621047