Learn More
Medchemexpress LLC HY-13704 500mg , SN-38 NK012 CAS:86639-52-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13704500MG
Encompass_Preferred
Medchemexpress, HY-13704 500mg SN-38 NK012 CAS:86639-52-3 Formula:C22H20N2O5 Topoisomerase I Purity:98% SN-38 is an active metabolite of the Topoisomerase I inhibitor Irinotecan. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2422