Learn More
Medchemexpress LLC SN-38-d3 | 718612-49-8 | MFCD09841270 | 98.8% | 395.42 | C22H17D3N2O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13704S5MG
Encompass_Preferred
SN-38-d3 is the deuterium-labeled analogue of SN-38, the active metabolite of irinotecan. Supplied as a high-purity stable isotope standard, it is intended for use as a tracer or internal standard in quantitative analytical methods and preclinical studies involving topoisomerase I activity.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 98.8%.
  • Suitable for NMR, GC-MS, and LC-MS workflows.
  • Useful as a tracer or internal standard in ADC payload and topoisomerase I research.
  • Available in small analytical pack sizes for laboratory use.
Catalog No. 50-002-22467