Learn More
Medchemexpress LLC SN-38 glucuronide | 121080-63-5 | 99.49% | 568.53 | 50 MG
SDP
Catalog No. 5000438073
Encompass_Preferred
SN-38 glucuronide is an inactive metabolite of the anticancer active molecule Irinotecan and has toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor used for researching colon and rectal cancer. SN38-G is the glucuronidated metabolite of SN38. Cells with the ability to metabolize SN38 to SN38-G are more resistant to extracellular SN38 than cells lacking this capability. SN-38 glucuronide can also serve as a substrate for gut microbial β-glucuronidase enzymes, which remove the glucuronic acid moiety to reactivate SN-38, thereby increasing its toxic effects on the gut.
  • For research use only.
Catalog No. 50-004-38073 Supplier Medchemexpress LLC Supplier No. HY12637350MG
SN-38 glucuronide is an inactive metabolite of the anticancer active molecule Irinotecan and has toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor used for researching colon and rectal cancer. SN38-G is the glucuronidated metabolite of SN38. Cells with the ability to metabolize SN38 to SN38-G are more resistant to extracellular SN38 than cells lacking this capability. SN-38 glucuronide can also serve as a substrate for gut microbial β-glucuronidase enzymes, which remove the glucuronic acid moiety to reactivate SN-38, thereby increasing its toxic effects on the gut.
  • For research use only.