Learn More
Medchemexpress LLC SN-38 glucuronide | 121080-63-5 | 99.5% | 568.53 | 1 MG
SDP
Catalog No. 5000438190
Encompass_Preferred
SN-38 glucuronide, also known as SN-38G, is an inactive metabolite of the anticancer molecule Irinotecan. It has known toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor used in research for colon and rectal cancer. SN-38 glucuronide can be reactivated by gut microbial β-glucuronidase enzymes, which remove the glucuronic acid moiety, thus increasing its toxic effects on the intestine.
  • Inactive metabolite of Irinotecan.
  • Has toxic effects on the gastrointestinal tract.
  • Can be reactivated by gut microbial β-glucuronidase enzymes.
  • Relevant in the study of colon and rectal cancer.
  • Cells metabolizing SN38 to SN38-G show increased resistance to extracellular SN38.
Catalog No. 50-004-38190 Supplier Medchemexpress LLC Supplier No. HY1263731MG
SN-38 glucuronide, also known as SN-38G, is an inactive metabolite of the anticancer molecule Irinotecan. It has known toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor used in research for colon and rectal cancer. SN-38 glucuronide can be reactivated by gut microbial β-glucuronidase enzymes, which remove the glucuronic acid moiety, thus increasing its toxic effects on the intestine.
  • Inactive metabolite of Irinotecan.
  • Has toxic effects on the gastrointestinal tract.
  • Can be reactivated by gut microbial β-glucuronidase enzymes.
  • Relevant in the study of colon and rectal cancer.
  • Cells metabolizing SN38 to SN38-G show increased resistance to extracellular SN38.