Learn More
Medchemexpress LLC SN-38 glucuronide | 121080-63-5 | MFCD00872153 | 99.9% | 568.53 | C28H28N2O11 | 5 MG
SDP
Catalog No. 5000264513
Encompass_Preferred
SN-38 glucuronide is the glucuronidated, inactive metabolite of the anticancer agent irinotecan, supplied for research use to study drug metabolism, pharmacokinetics, and cellular resistance mechanisms. The substance is characterized by CAS 121080-63-5, molecular formula C28H28N2O11, and molecular weight 568.53.
  • High purity suitable for analytical and biological studies.
  • Useful for metabolism and pharmacokinetic assays.
  • Enables investigation of cellular resistance to SN-38.
  • Suitable as a reference standard for LC-MS/MS and HPLC methods.
  • Provided in defined small-mass pack sizes for laboratory use.
Catalog No. 50-002-64513 Supplier Medchemexpress LLC Supplier No. HY1263735MG
SN-38 glucuronide is the glucuronidated, inactive metabolite of the anticancer agent irinotecan, supplied for research use to study drug metabolism, pharmacokinetics, and cellular resistance mechanisms. The substance is characterized by CAS 121080-63-5, molecular formula C28H28N2O11, and molecular weight 568.53.
  • High purity suitable for analytical and biological studies.
  • Useful for metabolism and pharmacokinetic assays.
  • Enables investigation of cellular resistance to SN-38.
  • Suitable as a reference standard for LC-MS/MS and HPLC methods.
  • Provided in defined small-mass pack sizes for laboratory use.