Learn More
Medchemexpress LLC SN-38 glucuronide (SN-38G) | 121080-63-5 | 99.5% | 568.53 | 10 MG
SDP
Catalog No. 5000438072
Encompass_Preferred
SN-38 glucuronide is an inactive metabolite of the anticancer active molecule Irinotecan and has toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor which can be used for researching colon and rectal cancer.
  • Inactive metabolite of the anticancer active molecule irinotecan.
  • Has toxic effects on the gastrointestinal tract.
  • Cells capable of metabolizing SN38 to SN38-G show increased resistance to extracellular SN38.
  • Can serve as a substrate for gut microbial β-glucuronidase enzymes, which reactivate SN-38 in the intestine, leading to increased toxic effects on the gut.
Catalog No. 50-004-38072 Supplier Medchemexpress LLC Supplier No. HY12637310MG
SN-38 glucuronide is an inactive metabolite of the anticancer active molecule Irinotecan and has toxic effects on the gastrointestinal tract. Irinotecan is a topoisomerase I inhibitor which can be used for researching colon and rectal cancer.
  • Inactive metabolite of the anticancer active molecule irinotecan.
  • Has toxic effects on the gastrointestinal tract.
  • Cells capable of metabolizing SN38 to SN38-G show increased resistance to extracellular SN38.
  • Can serve as a substrate for gut microbial β-glucuronidase enzymes, which reactivate SN-38 in the intestine, leading to increased toxic effects on the gut.