Learn More
Selleck Chemical LLC SN 52 P1238-25MG
SDP
Catalog No. 5000039363
Encompass_Preferred
SN52 a cell-permeable peptide containing the p50 NLS is a selective inhibitor of NF- B pathway It selectively and completely blocks the ionizing radiation (IR)-induced nuclear import of p52 and RelB It also enhances the radiosensitivity of prostate cancer cells
Catalog No. 50-000-39363 Supplier Selleck Chemical LLC Supplier No. P123825MG
SN52 a cell-permeable peptide containing the p50 NLS is a selective inhibitor of NF- B pathway It selectively and completely blocks the ionizing radiation (IR)-induced nuclear import of p52 and RelB It also enhances the radiosensitivity of prostate cancer cells